211280-100CM
211280-25CM
211280-500CM
BT211280-100CMEA
245.3
USD
InStock
BT211280-100CM
BT211280-25CM
BT211280-500CM
Gold ≥99.999% (trace metals basis), wire Ø 0.25 mm
Gold
CAS: 7440-57-5; EC No: 231-165-9; MDL No: MFCD00003436; RTECS: MD5420000
Solid; Molecular Formula: Au; MW: 196.97
Melting Point: 1063°; Boiling Point: 2808°
Density (g/mL): 19.3