Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Potassium chloride 3 M in water

Supplier: BEANTOWN CHEMICAL - BTC

211310-1L 211310-250ML
BT211310-1LEA 80.05 USD InStock
BT211310-1L BT211310-250ML
Potassium chloride 3 M in water
Potassium chloride
CAS: 7447-40-7; EC No: 231-211-8; MDL No: MFCD00011360 Liquid; Linear Formula: KCl; MW: 74.55 Boiling Point: 100° Density (g/mL): 1.12 Light Sensitive

  • Light Sensitive
  • Liquid


Formula: KCl
MW: 74.55 g/mol
Boiling Pt: 1420
Melting Pt: 773
Density: 1.12 g/mL
MDL Number: MFCD00011360
CAS Number: 7447-40-7
EINECS: 231-211-8
Merck Index: 14,07621
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty