Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Cyano-3-fluorophenylboronic acid ≥97%

Supplier: BEANTOWN CHEMICAL - BTC

211350-1G 211350-25G 211350-5G
BT211350-1GEA 85 USD InStock
BT211350-1G BT211350-25G BT211350-5G
4-Cyano-3-fluorophenylboronic acid ≥97%
4-Cyano-3-fluorophenylboronic acid
CAS: 843663-18-3; MDL No: MFCD03411549 Powder; Molecular Formula: C7H5BFNO2; MW: 164.93

  • Powder


Formula: C₇H₅BFNO₂
MW: 164.93 g/mol
MDL Number: MFCD03411549
CAS Number: 843663-18-3
Order Now
Specification Test Results
Assay 97%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty