Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl acetopyruvate ≥98%

Supplier: BEANTOWN CHEMICAL - BTC

211415-100G 211415-25G 211415-5G
BT211415-100GEA 623 USD InStock
BT211415-100G BT211415-25G BT211415-5G
Ethyl acetopyruvate ≥98%
Ethyl acetopyruvate
CAS: 615-79-2; EC No: 210-447-5; MDL No: MFCD00009124 Liquid; Linear Formula: CH3COCH2COCOOCH2CH3; Molecular Formula: C7H10O4; MW: 158.15 Melting Point: 16-18°; Boiling Point: 101-103°/12 mmHg; Flash point: 110°C (230°F) Density (g/mL): 1.126; Refractive Index: 1.470

  • Refractive index: 1.470
  • Liquid


Formula: C₇H₁₀O₄
Boiling Pt: 101…103 °C/12 mmHg
Melting Pt: 16…18 °C
Density: 1.126 g/mL
Flash Pt: >110 °C (>230 °F)
MDL Number: MFCD00009124
CAS Number: 615-79-2
EINECS: 210-447-5
Order Now
Specification Test Results
Assay 98%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty