Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-tert-Butylcalix[4]arene ≥99%

Supplier: BEANTOWN CHEMICAL - BTC

211440-25G 211440-5G
BT211440-25GEA 341.28 USD InStock
BT211440-25G BT211440-5G
4-tert-Butylcalix[4]arene ≥99%
4-tert-Butylcalix[4]arene
CAS: 60705-62-6; MDL No: MFCD00066280 Crystalline; Molecular Formula: C44H56O4 ; MW: 648.91 Melting Point: 300°

  • Crystalline


Formula: C₄₄H₅₆O₄
MW: 648.93 g/mol
Melting Pt: >300 °C
MDL Number: MFCD00066280
CAS Number: 60705-62-6
Order Now
Specification Test Results
Assay 99%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty