Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Cobalt ≥99.95% (trace metals basis), slug, Length: 6.35 mm, Diameter 3.175 mm

Supplier: BEANTOWN CHEMICAL - BTC

211950-100G 211950-25G 211950-5G
BT211950-100GEA 1025 USD InStock
BT211950-100G BT211950-25G BT211950-5G
Cobalt ≥99.95% (trace metals basis), slug, Length: 6.35 mm, Diameter 3.175 mm
Cobalt
CAS: 7440-48-4; EC No: 231-158-0; MDL No: MFCD00010935; RTECS: GF8750000 Solid; Molecular Formula: Co; MW: 58.93 Melting Point: 1495°; Boiling Point: 2900° Density (g/mL): 8.92

  • RTECS: GF8750000
  • Solid


Formula: Co
Boiling Pt: 2900 °C
Melting Pt: 1495 °C
Density: 8.92 g/mL
MDL Number: MFCD00010935
CAS Number: 7440-48-4
EINECS: 231-158-0
Order Now
Specification Test Results
Assay 99.95%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty