Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Iododecane ≥98% stabilised

Supplier: BEANTOWN CHEMICAL - BTC

212150-250G 212150-50G
BT212150-250GEA 262.9 USD InStock
BT212150-250G BT212150-50G
1-Iododecane ≥98% stabilised
1-Iododecane
CAS: 2050-77-3; EC No: 218-104-1; MDL No: MFCD00001085; RTECS: HD8705000 Liquid; Linear Formula: CH3(CH2)9I; Molecular Formula: C10H21I; MW: 268.18 Boiling Point: 132°/15 mmHg; Flash point: 110°C (230°F) Density (g/mL): 1.257; Refractive Index: 1.485 Light Sensitive

  • RTECS: HD8705000
  • Refractive index: 1.485
  • Light Sensitive
  • Liquid


Formula: C₁₀H₂₁I
Boiling Pt: 132 °C/15 mmHg
Density: 1.257 g/mL
Flash Pt: 110 °C (230 °F)
MDL Number: MFCD00001085
CAS Number: 2050-77-3
EINECS: 218-104-1
Order Now
Specification Test Results
Assay 98%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty