Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Aluminium sulphate hydrate 98.0-102.0% ACS

Supplier: BEANTOWN CHEMICAL - BTC

212360-1KG 212360-250G
BT212360-1KGEA 195.09 USD InStock
BT212360-1KG BT212360-250G
Aluminium sulphate hydrate 98.0-102.0% ACS
Aluminium sulphate hydrate
CAS: 17927-65-0; EC No: 233-135-0; MDL No: MFCD00149136; RTECS: BD1700000 Crystalline; Linear Formula: Al2(SO4)3·xH2O ; MW: 342.15 (anhydrous) Density (g/mL): 1.69

  • RTECS: BD1700000
  • Crystalline


Formula: Al₂(SO₄)₃·xH₂O
MW: 366.21 g/mol
Density: 1.69 g/mL
MDL Number: MFCD00149136
CAS Number: 17927-65-0
EINECS: 233-135-0
Order Now
Specification Test Results
Assay 98.0-102.0%

 

ORDER

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty