Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromo-3-nitro-2-picoline

Supplier: BEANTOWN CHEMICAL - BTC

212600-1G 212600-250MG
BT212600-1GEA 419 USD InStock
BT212600-1G BT212600-250MG
5-Bromo-3-nitro-2-picoline
5-Bromo-3-nitro-2-picoline
CAS: 911434-05-4; MDL No: MFCD09031419 Solid; Molecular Formula: C6H5BrN2O2 ; MW: 217.02 Density (g/mL): 1.709

  • Solid


Formula: C₆H₅BrN₂O₂
MW: 217.02
Density: 1.709 g/mL
MDL Number: MFCD09031419
CAS Number: 911434-05-4
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty