Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Isatin ≥98%

Supplier: BEANTOWN CHEMICAL - BTC

212780-100G 212780-2.5KG 212780-500G
BT212780-100GEA 24 USD InStock
BT212780-100G BT212780-2.5KG BT212780-500G
Isatin ≥98%
Isatin
CAS: 91-56-5; EC No: 202-077-8; MDL No: MFCD00005718; RTECS: NL7873000 Liquid; Molecular Formula: C8H5NO2; MW: 147.13 Melting Point: 193-195° (decomposes); Flash point: 220°C (428°F)

  • RTECS: NL7873000
  • Liquid


Formula: C₈H₅NO₂
MW: 147.13 g/mol
Boiling Pt: 342
Melting Pt: 193…195 °C (dec.)
Flash Pt: 220 °C (428 °F)
MDL Number: MFCD00005718
CAS Number: 91-56-5
EINECS: 202-077-8
Merck Index: 13,05119
Order Now
Specification Test Results
Assay 98%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty