Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Nitrophenyl octyl ether ≥98%

Supplier: BEANTOWN CHEMICAL - BTC

212820-25G 212820-5G
BT212820-25GEA 212 USD InStock
BT212820-25G BT212820-5G
2-Nitrophenyl octyl ether ≥98%
2-Nitrophenyl octyl ether
CAS: 37682-29-4; EC No: 253-623-7; MDL No: MFCD00014693 Liquid; Molecular Formula: C14H21NO3; MW: 251.32 Boiling Point: 197-198°/11 mmHg; Flash point: 110°C (230°F) Density (g/mL): 1.04; Refractive Index: 1.508

  • Refractive index: 1.508
  • Liquid


Formula: C₁₄H₂₁NO₃
Boiling Pt: 197…198 °C/11 mmHg
Density: 1.04 g/mL
Flash Pt: >110 °C (>230 °F)
MDL Number: MFCD00014693
CAS Number: 37682-29-4
EINECS: 253-623-7
Order Now
Specification Test Results
Assay 98%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty