Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,7-Heptanediol ≥98%

Supplier: BEANTOWN CHEMICAL - BTC

212955-25G 212955-5G
BT212955-25GEA 182 USD InStock
BT212955-25G BT212955-5G
1,7-Heptanediol ≥98%
1,7-Heptanediol
CAS: 629-30-1; EC No: 211-085-0; MDL No: MFCD00002987; RTECS: MI9804000 Liquid; Linear Formula: HO(CH2)7OH; Molecular Formula: C7H16O2; MW: 132.20 Melting Point: 17-19°; Boiling Point: 259°; Flash point: 110°C (230°F) Density (g/mL): 0.951; Refractive Index: 1.455

  • RTECS: MI9804000
  • Refractive index: 1.455
  • Liquid


Formula: C₇H₁₆O₂
Boiling Pt: 259 °C
Melting Pt: 17…19 °C
Density: 0.951 g/mL
Flash Pt: >110 °C (>230 °F)
MDL Number: MFCD00002987
CAS Number: 629-30-1
EINECS: 211-085-0
Order Now
Specification Test Results
Assay 98%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty