Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Triphenylphosphine ≥99%

Supplier: BEANTOWN CHEMICAL - BTC

213420-100G 213420-25G 213420-500G
BT213420-100GEA 55.4 USD InStock
BT213420-100G BT213420-25G BT213420-500G
Triphenylphosphine ≥99%
Triphenylphosphine
CAS: 603-35-0; EC No: 210-036-0; MDL No: MFCD00003043; RTECS: SZ3500000 Crystalline; Linear Formula: (C6H5)3P; Molecular Formula: C18H15P; MW: 262.29 Melting Point: 79-81°; Boiling Point: 377°; Flash point: 181°C (358°F) Density (g/mL): 1.2

  • RTECS: SZ3500000
  • Crystalline


Formula: C₁₈H₁₅P
Boiling Pt: 377 °C
Melting Pt: 79…81 °C
Density: 1.2 g/mL
Flash Pt: 181 °C (358 °F)
MDL Number: MFCD00003043
CAS Number: 603-35-0
EINECS: 210-036-0
Order Now
Specification Test Results
Assay 99%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty