Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tantalum ≥99.95% (trace metals basis), rods Ø 12.7 mm

Supplier: BEANTOWN CHEMICAL - BTC

213525-1CM 213525-25CM 213525-5CM
BT213525-1CMEA 117 USD InStock
BT213525-1CM BT213525-25CM BT213525-5CM
Tantalum ≥99.95% (trace metals basis), rods Ø 12.7 mm
Tantalum
CAS: 7440-25-7; EC No: 231-135-5; MDL No: MFCD00011252 Solid; Molecular Formula: Ta; MW: 180.95 Melting Point: 2996°; Boiling Point: 5425° Density (g/mL): 16.69

  • Solid


Formula: Ta
MW: 180.95 g/mol
Boiling Pt: 5427
Melting Pt: 2996
Density: 16.69 g/mL
MDL Number: MFCD00011252
CAS Number: 7440-25-7
EINECS: 231-135-5
Merck Index: 12,09223
Order Now
Specification Test Results
Assay 99.95%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty