Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tungstic(VI) acid

Supplier: BEANTOWN CHEMICAL - BTC

213800-100G 213800-25G 213800-500G
BT213800-100GEA 74.84 USD InStock
BT213800-100G BT213800-25G BT213800-500G
Tungstic(VI) acid
Tungstic(VI) acid
CAS: 7783-03-1; EC No: 231-975-2; MDL No: MFCD00011336 Powder; Linear Formula: H2WO4; MW: 249.86 Melting Point: 100° (decomposes); Boiling Point: 1473° Density (g/mL): 5.5

  • Powder


Formula: H₂O₄W
Boiling Pt: 1473 °C
Melting Pt: 100 °C (dec.)
Density: 5.5 g/mL
MDL Number: MFCD00011336
CAS Number: 7783-03-1
EINECS: 231-975-2
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty