Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Gold ≥99.95% (trace metals basis), powder ≤10 µm

Supplier: BEANTOWN CHEMICAL - BTC

213900-1G 213900-5G
BT213900-1GEA 196 USD InStock
BT213900-1G BT213900-5G
Gold ≥99.95% (trace metals basis), powder ≤10 µm
Gold
CAS: 7440-57-5; EC No: 231-165-9; MDL No: MFCD00003436 Powder, S.A. 0.35-0.75 m2/g; Molecular Formula: Au; MW: 196.97 Melting Point: 1063°; Boiling Point: 2808° Density (g/mL): 19.3

  • Powder, S.A. 0.35-0.75 m²/g


Formula: Au
MW: 196.97 g/mol
Boiling Pt: 2800
Melting Pt: 1063
Density: 19.3 g/mL
MDL Number: MFCD00003436
CAS Number: 7440-57-5
EINECS: 231-165-9
Merck Index: 12,04526
Order Now
Specification Test Results
Assay 99.95%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty