Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Acetyladamantane ≥99%

Supplier: BEANTOWN CHEMICAL - BTC

214335-1G 214335-25G 214335-5G
BT214335-1GEA 33 USD InStock
BT214335-1G BT214335-25G BT214335-5G
1-Acetyladamantane ≥99%
1-Acetyladamantane
CAS: 1660-04-4; EC No: 216-761-9; MDL No: MFCD00074739 Crystalline/Powder; Molecular Formula: C12H18O; MW: 178.27 Melting Point: 53-55°; Flash point: 108°C (226°F)

  • Crystalline/Powder


Formula: C₁₂H₁₈O
Melting Pt: 53…55 °C
Flash Pt: 108 °C (226 °F)
MDL Number: MFCD00074739
CAS Number: 1660-04-4
EINECS: 216-761-9
Order Now
Specification Test Results
Assay 99%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty