Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Zinc ≥99.999% (trace metals basis), foil, Thickness: 2 mm

Supplier: BEANTOWN CHEMICAL - BTC

214355-25X25MM 214355-50X50MM
BT214355-25X25EA 493 USD InStock
BT214355-25X25 BT214355-50X50
Zinc ≥99.999% (trace metals basis), foil, Thickness: 2 mm
Zinc
CAS: 7440-66-6; EC No: 231-175-3; MDL No: MFCD00011291 Solid; Molecular Formula: Zn; MW: 65.39 Melting Point: 420°; Boiling Point: 907° Density (g/mL): 7.133

  • Solid


Formula: Zn
MW: 65.39 g/mol
Boiling Pt: 908
Melting Pt: 420
Density: 7.133 g/mL
MDL Number: MFCD00011291
CAS Number: 7440-66-6
EINECS: 231-175-3
Merck Index: 12,10255
Order Now
Specification Test Results
Assay 99.999%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty