Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-tert-Butyl-5-oxopyrrolidine-2-carboxylate ≥95%

Supplier: BEANTOWN CHEMICAL - BTC

214360-25G 214360-5G
BT214360-25GEA 241 USD InStock
BT214360-25G BT214360-5G
(S)-tert-Butyl-5-oxopyrrolidine-2-carboxylate ≥95%
(S)-tert-Butyl-5-oxopyrrolidine-2-carboxylate
CAS: 35418-16-7; EC No: 252-555-5; MDL No: MFCD06659481 Powder; Molecular Formula: C9H15NO3; MW: 185.22 Melting Point: 91-92°

  • Powder


Formula: C₉H₁₅NO₃
MW: 185.22 g/mol
Melting Pt: 91…92 °C
MDL Number: MFCD06659481
CAS Number: 35418-16-7
EINECS: 252-555-5
Order Now
Specification Test Results
Assay 95%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty