Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4-Diaminoquinazoline ≥98%

Supplier: BEANTOWN CHEMICAL - BTC

214425-1G 214425-5G
BT214425-1GEA 37 USD InStock
BT214425-1G BT214425-5G
2,4-Diaminoquinazoline ≥98%
2,4-Diaminoquinazoline
CAS: 1899-48-5; MDL No: MFCD00236032 Solid; Molecular Formula: C8H8N4; MW: 160.18

  • Solid


Formula: C₈H₈N₄
MDL Number: MFCD00236032
CAS Number: 1899-48-5
Order Now
Specification Test Results
Assay 98%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty