214475-1G
214475-25G
214475-5G
BT214475-1GEA
23
USD
InStock
BT214475-1G
BT214475-25G
BT214475-5G
2,5-Thiophenedicarboxylic acid ≥97%
2,5-Thiophenedicarboxylic acid
CAS: 4282-31-9; EC No: 224-284-2; MDL No: MFCD00016896
Powder; Molecular Formula: C6H4O4S; MW: 172.16
Melting Point: 355-360°