Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Poly(styrene sulfonic acid), M.W. ∼75000

Supplier: BEANTOWN CHEMICAL - BTC

214545-100G 214545-500G
BT214545-100GEA 63.2 USD InStock
BT214545-100G BT214545-500G
Poly(styrene sulfonic acid), M.W. ∼75000
Poly(styrene sulfonic acid)
CAS: 28210-41-5; MDL No: MFCD00165973 Liquid; Linear Formula: [-CH2CH(C6H4SO3H)-]n Density (g/mL): 1.1

  • Liquid


Formula: [-CH₂CH(C₆H₄SO₃H)-]n
Density: 1.1 g/mL
MDL Number: MFCD00165973
CAS Number: 28210-41-5
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty