Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Methyl-1-nonyn-3-ol ≥97%

Supplier: BEANTOWN CHEMICAL - BTC

214850-100G 214850-25G 214850-5G
BT214850-100GEA 356 USD InStock
BT214850-100G BT214850-25G BT214850-5G
3-Methyl-1-nonyn-3-ol ≥97%
3-Methyl-1-nonyn-3-ol
CAS: 5430-01-3; MDL No: MFCD00041582 Liquid; Molecular Formula: C10H18O; MW: 154.25 Boiling Point: 87-88°/15 mmHg

  • Liquid


Formula: C₁₀H₁₈O
Boiling Pt: 87…88 °C/15 mmHg
MDL Number: MFCD00041582
CAS Number: 5430-01-3
Order Now
Specification Test Results
Assay 97%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty