Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Bentonite

Supplier: BEANTOWN CHEMICAL - BTC

215135-2.5KG 215135-500G
BT215135-2.5KGEA 86.45 USD InStock
BT215135-2.5KG BT215135-500G
Bentonite
Bentonite
CAS: 1302-78-9; EC No: 215-108-5; MDL No: MFCD00130611 Granular

  • Granular


MDL Number: MFCD00130611
CAS Number: 1302-78-9
EINECS: 215-108-5
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty