Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Methyl-2-nitrobenzoic acid ≥98%

Supplier: BEANTOWN CHEMICAL - BTC

215180-100G 215180-25G
BT215180-100GEA 149 USD InStock
BT215180-100G BT215180-25G
3-Methyl-2-nitrobenzoic acid ≥98%
3-Methyl-2-nitrobenzoic acid
CAS: 5437-38-7; EC No: 226-610-9; MDL No: MFCD00007180 Crystalline/Powder; Linear Formula: C6H3(CH3)(NO2)COOH; Molecular Formula: C8H7NO4; MW: 181.15 Melting Point: 219-223°

  • Crystalline/Powder


Formula: C₈H₇NO₄
MW: 181.15 g/mol
Boiling Pt: 340
Melting Pt: 219…223 °C
MDL Number: MFCD00007180
CAS Number: 5437-38-7
Order Now
Specification Test Results
Assay 98%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty