Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

≥98.0% ACS

Supplier: BEANTOWN CHEMICAL - BTC

215585-1KG 215585-250G 215585-5KG
BT215585-1KGEA 202.16 USD InStock
BT215585-1KG BT215585-250G BT215585-5KG
≥98.0% ACS
di-Potassium hydrogen phosphate
CAS: 7758-11-4; EC No: 231-834-5; MDL No: MFCD00011383; RTECS: TC5580000 Granular; Linear Formula: K2HPO4; MW: 174.18 Melting Point: 340° Density (g/mL): 2.3 Hygroscopic

  • RTECS: TC5580000
  • Hygroscopic
  • Granular


Formula: K₂HPO₄
MW: 174.18 g/mol
Melting Pt: 340
Density: 2.3 g/mL
MDL Number: MFCD00011383
CAS Number: 7758-11-4
EINECS: 231-834-5
Merck Index: 13,07743
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ee2dc83ce0381f3a1e1f9b57dbc5d1d8ba841eaa&vid=bf7c831e84a9e041f3acaa70912b0ef870c7ea23&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Assay 98.0% min.
pH of 5% solution at 25°C 8.5-9.6
Insoluble matter ≤0.01%
Loss on drying (105°C) ≤1.0%
Chloride (Cl) ≤0.003%
Nitrogen compounds (as N) ≤0.001%
Sulfate (SO4) ≤0.005%
Heavy metals (as Pb) ≤5ppm
Iron (Fe) ≤0.001%
Sodium (Na) ≤0.05%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty