Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Water for HPLC

Supplier: BEANTOWN CHEMICAL - BTC

215795-1L 215795-4L 215795-4X4L
BT215795-1LEA 49.31 USD InStock
BT215795-1L BT215795-4L BT215795-4X4L
Water for HPLC
Water
CAS: 7732-18-5; EC No: 231-791-2; MDL No: MFCD00011332 Liquid; Molecular Formula: H2O ; MW: 18.02 Melting Point: 0°; Boiling Point: 100° Density (g/mL): 1; Refractive Index: 1.333


Formula: H₂O
MW: 18.02 g/mol
Boiling Pt: 100
Melting Pt: 0
Density: 1 g/mL
MDL Number: MFCD00011332
CAS Number: 7732-18-5
EINECS: 231-791-2
Merck Index: 13,10098
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty