Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Amino-4-bromopyridine ≥97%

Supplier: BEANTOWN CHEMICAL - BTC

215955-1G 215955-250MG
BT215955-1GEA 262 USD InStock
BT215955-1G BT215955-250MG
2-Amino-4-bromopyridine ≥97%
2-Amino-4-bromopyridine
CAS: 84249-14-9; MDL No: MFCD01646115 Solid; Molecular Formula: C5H5BrN2; MW: 173.01 Melting Point: 142-146°

  • Solid


Formula: C₅H₅BrN₂
MW: 173.01
Melting Pt: 142…146 °C
MDL Number: MFCD01646115
CAS Number: 84249-14-9
Order Now
Specification Test Results
Assay 97%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty