Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Glycine-tert-butyl ester(H-Gly-OtBu) ≥97%

Supplier: BEANTOWN CHEMICAL - BTC

216475-1G 216475-5G
BT216475-1GEA 65 USD InStock
BT216475-1G BT216475-5G
Glycine-tert-butyl ester(H-Gly-OtBu) ≥97%
Glycine-tert-butyl ester(H-Gly-OtBu)
CAS: 6456-74-2; MDL No: MFCD00038194 Liquid; Linear Formula: H2NCH2CO2C(CH3)3; Molecular Formula: C6H13NO2; MW: 131.18 Boiling Point: 29-31°/2 mmHg Density (g/mL): 0.96 Air Sensitive

  • Air Sensitive
  • Liquid


Formula: C₆H₁₃NO₂
MW: 131.18 g/mol
Boiling Pt: 29…31 °C/2 mmHg
Density: 0.96 g/mL
MDL Number: MFCD00038194
CAS Number: 6456-74-2
Order Now
Specification Test Results
Assay 97%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty