Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Methylthio-4-pyrimidinol ≥98%

Supplier: BEANTOWN CHEMICAL - BTC

216645-1G 216645-25G 216645-5G
BT216645-1GEA 34 USD InStock
BT216645-1G BT216645-25G BT216645-5G
2-Methylthio-4-pyrimidinol ≥98%
2-Methylthio-4-pyrimidinol
CAS: 5751-20-2; EC No: 227-274-6; MDL No: MFCD00047373 Solid; Molecular Formula: C5H6N2OS; MW: 142.18

  • Solid


Formula: C₅H₆N₂OS
MW: 142.18 g/mol
MDL Number: MFCD00047373
CAS Number: 5751-20-2
EINECS: 227-274-6
Order Now
Specification Test Results
Assay 98%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty