216685-100G
216685-25G
BT216685-100GEA
206
USD
InStock
BT216685-100G
BT216685-25G
Copper ≥99.9999% (trace metals basis), slug, Length: 12.7 mm, Diameter 6.35 mm
Copper
CAS: 7440-50-8; EC No: 231-159-6; MDL No: MFCD00010965; RTECS: GL5325000
Solid; Molecular Formula: Cu; MW: 63.55
Melting Point: 1083.4°; Boiling Point: 2567°
Density (g/mL): 8.94