Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Nickel ≥99.95% (trace metals basis excluding Co), foil, not supported, Thickness: 1.0 µm

Supplier: BEANTOWN CHEMICAL - BTC

216690-25X25MM 216690-50X50MM
BT216690-25X25EA 371 USD InStock
BT216690-25X25 BT216690-50X50
Nickel ≥99.95% (trace metals basis excluding Co), foil, not supported, Thickness: 1.0 µm
Nickel
CAS: 7440-02-0; EC No: 231-111-4; MDL No: MFCD00011137 Solid; Molecular Formula: Ni; MW: 58.69 Melting Point: 1453°; Boiling Point: 2732° Density (g/mL): 8.9

  • Solid


Formula: Ni
Boiling Pt: 2732 °C
Melting Pt: 1453 °C
Density: 8.9 g/mL
MDL Number: MFCD00011137
CAS Number: 7440-02-0
EINECS: 231-111-4
UN: 3089
ADR: 4.1,III
Order Now
Specification Test Results
Assay 99.95%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty