Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1H-Pyrazole ≥98%

Supplier: BEANTOWN CHEMICAL - BTC

217075-100G 217075-25G
BT217075-100GEA 141 USD InStock
BT217075-100G BT217075-25G
1H-Pyrazole ≥98%
1H-Pyrazole
CAS: 288-13-1; EC No: 206-017-1; MDL No: MFCD00005234; RTECS: UQ4900000 Crystalline; Molecular Formula: C3H4N2; MW: 68.08 Melting Point: 67-70°; Boiling Point: 186-188° Hygroscopic

  • RTECS: UQ4900000
  • Hygroscopic
  • Crystalline


Formula: C₃H₄N₂
MW: 68.08 g/mol
Boiling Pt: 186
Melting Pt: 67…70 °C
MDL Number: MFCD00005234
CAS Number: 288-13-1
EINECS: 206-017-1
Merck Index: 12,08144
Order Now
Specification Test Results
Assay 98%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty