Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Bis(ethylenedithio)tetrathiafulvalene

Supplier: BEANTOWN CHEMICAL - BTC

217225-1G 217225-250MG
BT217225-1GEA 306 USD InStock
BT217225-1G BT217225-250MG
Bis(ethylenedithio)tetrathiafulvalene
Bis(ethylenedithio)tetrathiafulvalene
CAS: 66946-48-3; MDL No: MFCD00059710 Powder; Molecular Formula: C10H8S8; MW: 384.68 Melting Point: 238° (decomposes)

  • Powder


Formula: C₁₀H₈S₈
Melting Pt: 238 °C (dec.)
MDL Number: MFCD00059710
CAS Number: 66946-48-3
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty