Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Aluminium ≥99.999% (trace metals basis), wire Ø 1.0 mm, annealed

Supplier: BEANTOWN CHEMICAL - BTC

217275-100M 217275-25M 217275-5M
BT217275-100MEA 476 USD InStock
BT217275-100M BT217275-25M BT217275-5M
Aluminium ≥99.999% (trace metals basis), wire Ø 1.0 mm, annealed
Aluminium
CAS: 7429-90-5; EC No: 231-072-3; MDL No: MFCD00134029; RTECS: BD0330006 Solid; Molecular Formula: Al; MW: 26.98 Melting Point: 660.4°; Boiling Point: 2460° Density (g/mL): 2.7

  • RTECS: BD0330006
  • Solid


Formula: Al
MW: 26.98 g/mol
Boiling Pt: 2327
Melting Pt: 660
Density: 2.7 g/mL
MDL Number: MFCD00134029
CAS Number: 7429-90-5
EINECS: 231-072-3
Merck Index: 13,00321
Order Now
Specification Test Results
Assay 99.999%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty