Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,14-Tetradecanedioic acid

Supplier: BEANTOWN CHEMICAL - BTC

217490-25G 217490-5G
BT217490-25GEA 224 USD InStock
BT217490-25G BT217490-5G
1,14-Tetradecanedioic acid
1,14-Tetradecanedioic acid
CAS: 821-38-5; EC No: 212-476-9; MDL No: MFCD00002743 Powder; Linear Formula: HOOC(CH2)12COOH; Molecular Formula: C14H26O4 ; MW: 258.35 Melting Point: 124-127°

  • Powder


Formula: C₁₄H₂₆O₄
Melting Pt: 124…127 °C
MDL Number: MFCD00002743
CAS Number: 821-38-5
EINECS: 212-476-9
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty