Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Magnesium silicate for chromatography

Supplier: BEANTOWN CHEMICAL - BTC

217520-10G 217520-250G 217520-50G
BT217520-10GEA 70 USD InStock
BT217520-10G BT217520-250G BT217520-50G
Magnesium silicate for chromatography
Magnesium silicate
CAS: 1343-88-0; EC No: 215-681-1; MDL No: MFCD00078249 60-100 Mesh Powder; Molecular Formula: Mg2O4Si ; MW: 140.71 Melting Point: 1910° Density (g/mL): 0.127

  • 60-100 Mesh Powder


Formula: MgO₃Si
MW: 100.38 g/mol
Melting Pt: 1910 °C
Density: 0.127 g/mL
MDL Number: MFCD00078249
CAS Number: 1343-88-0
EINECS: 215-681-1
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty