Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Cerium(III) carbonate hydrate ≥99.9%

Supplier: BEANTOWN CHEMICAL - BTC

217625-100G 217625-500G
BT217625-100GEA 74 USD InStock
BT217625-100G BT217625-500G
Cerium(III) carbonate hydrate ≥99.9%
Cerium (III) carbonate hydrate
CAS: 54451-25-1; EC No: 208-655-6; MDL No: MFCD00149635 Crystalline; Linear Formula: Ce2(CO3)3·xH2O; MW: 460.27 (anhydrous) Hygroscopic

  • Hygroscopic
  • Crystalline


Formula: C₃Ce₂O₉·H₂O
MW: 478.274 g/mol
MDL Number: MFCD00149635
CAS Number: 54451-25-1
EINECS: 208-655-6
Order Now
Specification Test Results
Assay 99.9%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty