Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Chalcopyrite naturally occurring mineral, grains, Diameter ∼ 1.5-4.8 mm

Supplier: BEANTOWN CHEMICAL - BTC

217715-1G 217715-25G 217715-5G
BT217715-1GEA 24 USD InStock
BT217715-1G BT217715-25G BT217715-5G
Chalcopyrite naturally occurring mineral, grains, Diameter ∼ 1.5-4.8 mm
Chalcopyrite
CAS: 1308-56-1; MDL No: MFCD00064817 Granular

  • Granular


Formula: CuFeH₄S₂
MDL Number: MFCD00064817
CAS Number: 1308-56-1
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty