Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromopyridinium chloride ≥99%

Supplier: BEANTOWN CHEMICAL - BTC

218605-1G 218605-25G 218605-5G
BT218605-1GEA 34 USD InStock
BT218605-1G BT218605-25G BT218605-5G
4-Bromopyridinium chloride ≥99%
4-Bromopyridinium chloride
CAS: 19524-06-2; EC No: 243-128-4; MDL No: MFCD00012828 Crystalline; Molecular Formula: C5H4BrN·HCl ; MW: 194.46 Melting Point: 270° (decomposes) Hygroscopic

  • Hygroscopic
  • Crystalline


Formula: C₅H₅BrClN
Melting Pt: 270 °C (dec.)
MDL Number: MFCD00012828
CAS Number: 19524-06-2
EINECS: 243-128-4
UN: 2811
ADR: 6.1,III
Order Now
Specification Test Results
Assay 99%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty