Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dodecylbenzene mixture of isomers

Supplier: BEANTOWN CHEMICAL - BTC

218620-1KG 218620-250G
BT218620-1KGEA 180 USD InStock
BT218620-1KG BT218620-250G
Dodecylbenzene mixture of isomers
Dodecylbenzene
CAS: 123-01-3; MDL No: MFCD00008974 Liquid; Linear Formula: C6H5(CH2)11CH3; Molecular Formula: C6H5C12H25; MW: 246.44 Melting Point: 3°; Boiling Point: 185-188°/15 mmHg Density (g/mL): 0.856; Refractive Index: 1.482

  • Refractive index: 1.482
  • Liquid


Formula: C₁₈H₃₀
MW: 246.44 g/mol
Boiling Pt: 275
Melting Pt: -4
Density: 0.856 g/mL
MDL Number: MFCD00008974
CAS Number: 123-01-3
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty