Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Silicon(II) oxide ≥99.5%

Supplier: BEANTOWN CHEMICAL - BTC

218680-250G 218680-50G
BT218680-250GEA 407 USD InStock
BT218680-250G BT218680-50G
Silicon(II) oxide ≥99.5%
Silicon(II) oxide
CAS: 10097-28-6; EC No: 233-232-8; MDL No: MFCD00151536 -325 Mesh Powder; Linear Formula: SiO; MW: 44.09 Melting Point: 1702°; Boiling Point: 1880° Density (g/mL): 2.13

  • -325 Mesh Powder


Formula: SiO
MW: 44.08 g/mol
Boiling Pt: 1880
Melting Pt: 1702
Density: 2.13 g/mL
MDL Number: MFCD00151536
CAS Number: 10097-28-6
EINECS: 233-232-8
Merck Index: 12,08640
Order Now
Specification Test Results
Assay 99.5%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty