Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sodium tungstate dihydrate ≥95%

Supplier: BEANTOWN CHEMICAL - BTC

218690-1KG 218690-250G
BT218690-1KGEA 211 USD InStock
BT218690-1KG BT218690-250G
Sodium tungstate dihydrate ≥95%
Sodium tungstate dihydrate
CAS: 10213-10-2; EC No: 236-743-4; MDL No: MFCD00149190; RTECS: YO7900000 Crystalline; Linear Formula: Na2WO4·2H2O ; MW: 329.86 Melting Point: 698° Density (g/mL): 4.18

  • RTECS: YO7900000
  • Crystalline


Formula: Na₂WO₄·2H₂O
MW: 329.85 g/mol
Melting Pt: 100
Density: 4.18 g/mL
MDL Number: MFCD00149190
CAS Number: 10213-10-2
EINECS: 236-743-4
Merck Index: 12,08847
Order Now
Specification Test Results
Assay 95%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty