Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

L(+)-Valine ≥99%

Supplier: BEANTOWN CHEMICAL - BTC

218745-100G 218745-25G 218745-500G
BT218745-100GEA 67.52 USD InStock
BT218745-100G BT218745-25G BT218745-500G
L(+)-Valine ≥99%
L(+)-Valine
CAS: 72-18-4; EC No: 200-773-6; MDL No: MFCD00064220; RTECS: YV9361000 Solid; Molecular Formula: C5H11NO2; MW: 117.15 Melting Point: 295-300° (sublimes) Optical Rotation: [α]20/D +27.0±0.5°, c = 5% in 5M HCl

  • RTECS: YV9361000
  • Optical rotation: [α]20/D +27.0±0.5°, c = 5% in 5M HCl
  • Solid


Formula: C₅H₁₁NO₂
MW: 117.15 g/mol
Boiling Pt: 217
Melting Pt: 86 (subl.)
MDL Number: MFCD00064220
CAS Number: 72-18-4
EINECS: 200-773-6
Merck Index: 13,09458
Order Now
Specification Test Results
Assay 99%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty