218775-1KG
218775-250G
218775-5KG
BT218775-1KGEA
101.21
USD
InStock
BT218775-1KG
BT218775-250G
BT218775-5KG
Disodium hydrogen phosphate, anhydrous ≥99.0% ACS
Disodium hydrogen phosphate
CAS: 7558-79-4; EC No: 231-448-7; MDL No: MFCD00003496; RTECS: WC4500000
Powder; Molecular Formula: HNa2O4P; MW: 141.98
Density (g/mL): 1.52
Hygroscopic
- RTECS: WC4500000
- Hygroscopic
- Powder