Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tellurium dioxide ≥99.99%

Supplier: BEANTOWN CHEMICAL - BTC

218820-100G 218820-25G
BT218820-100GEA 261.03 USD InStock
BT218820-100G BT218820-25G
Tellurium dioxide ≥99.99%
Tellurium dioxide
CAS: 7446-07-3; EC No: 231-193-1; MDL No: MFCD00011263; RTECS: WY2675000 Powder; Linear Formula: TeO2; MW: 159.60 Melting Point: 733° Density (g/mL): 5.67

  • RTECS: WY2675000
  • Powder


Formula: O₂Te
MW: 159.6 g/mol
Boiling Pt: 1245
Melting Pt: 773
Density: 5.67 g/mL
MDL Number: MFCD00011263
CAS Number: 7446-07-3
EINECS: 231-193-1
Merck Index: 14,09123
Order Now
Specification Test Results
Assay 99.99%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty