Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-(-)-N-Benzyl-α-methylbenzylamine ≥99%

Supplier: BEANTOWN CHEMICAL - BTC

219255-25G 219255-5G
BT219255-25GEA 124 USD InStock
BT219255-25G BT219255-5G
(S)-(-)-N-Benzyl-α-methylbenzylamine ≥99%
(S)-(-)-N-Benzyl-α-methylbenzylamine
CAS: 17480-69-2; MDL No: MFCD00066325 Liquid; Molecular Formula: C15H17N ; MW: 211.30 Boiling Point: 171°/15 mmHg; Flash point: 110°C (230°F) Density (g/mL): 1.01; Refractive Index: 1.563

  • Refractive index: 1.563
  • Liquid


Formula: C₁₅H₁₇N
Boiling Pt: 171 °C/15 mmHg
Density: 1.01 g/mL
Flash Pt: >110 °C (>230 °F)
MDL Number: MFCD00066325
CAS Number: 17480-69-2
Order Now
Specification Test Results
Assay 99%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty