Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

L(+)-Glutamic acid ≥99%

Supplier: BEANTOWN CHEMICAL - BTC

219400-1KG 219400-250G
BT219400-1KGEA 54.83 USD InStock
BT219400-1KG BT219400-250G
L(+)-Glutamic acid ≥99%
L(+)-Glutamic acid
CAS: 56-86-0; EC No: 200-293-7; MDL No: MFCD00002634; RTECS: LZ9700000 Crystalline; Molecular Formula: C5H9NO4; MW: 147.13 Melting Point: 205° (decomposes)

  • RTECS: LZ9700000
  • Crystalline


Formula: HO₂CCH₂CH₂CH(NH₂)CO₂H
Melting Pt: 205 °C (dec.)
MDL Number: MFCD00002634
CAS Number: 56-86-0
EINECS: 200-293-7
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e1cb2faefcc53483f0b3bfccd6c12b611e487241&vid=fdb021cd4468aa68a6e5186d44249b5e605f4aed&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Assay 99%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty