Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Muscovite (from natural source), sheets

Supplier: BEANTOWN CHEMICAL - BTC

219570-1PC 219570-5PC
BT219570-1PCEA 94 USD InStock
BT219570-1PC BT219570-5PC
Muscovite (from natural source), sheets
Muscovite
CAS: 12001-26-2; MDL No: MFCD00151447; RTECS: VV8760000 Sheet, apx 15cm x 15cm; Molecular Formula: K2O·3Al2O3·6O2Si·2H2O; MW: 796.65

  • RTECS: VV8760000
  • Sheet, apx 15cm x 15cm


Formula: K₂O.₃Al₂O₃.₆SiO₂•₂H₂O
MDL Number: MFCD00151447
CAS Number: 12001-26-2
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty