Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Hydroxy-7-methoxyquinoline-3-carboxylic acid ≥95%

Supplier: BEANTOWN CHEMICAL - BTC

219715-1G 219715-5G
BT219715-1GEA 47 USD InStock
BT219715-1G BT219715-5G
4-Hydroxy-7-methoxyquinoline-3-carboxylic acid ≥95%
4-Hydroxy-7-methoxyquinoline-3-carboxylic acid
CAS: 28027-17-0; MDL No: MFCD00024004 Solid; Molecular Formula: C11H9NO4; MW: 219.19

  • Solid


Formula: C₁₁H₉NO₄
MDL Number: MFCD00024004
CAS Number: 28027-17-0
Order Now
Specification Test Results
Assay 95%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty